missing translation for 'onlineSavingsMsg'
Learn More

6'-Amino-3',4'-(methylenedioxy)acetophenone 98.0+%, TCI America™

Catalog No. A13565G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A13565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A13565G Supplier TCI America Supplier No. A13565G
Only null left

Specifications

CAS 28657-75-2
Melting Point 171°C
Molecular Formula C9H9NO3
Quantity 5 g
MDL Number MFCD00075855
Synonym 1-6-aminobenzo d 1,3 dioxol-5-yl ethanone, 6'-amino-3',4'-methylenedioxy acetophenone, 1-6-amino-1,3-benzodioxol-5-yl ethanone, 2'-amino-4',5'-methylenedioxy acetophenone, 2-amino-4,5-methylenedioxyacetophenone, 1-6-amino-2h-1,3-benzodioxol-5-yl ethan-1-one, 1-6-amino-1,3-benzodioxol-5-yl ethan-1-one, 2'-amino-4',5'-methylenedioxyacetophenone, 5-acetyl-6-amino-1,3-benzodioxole, 1-6-amino-2h-1,3-benzodioxol-5-yl ethanone
InChI Key DWTHYSZSRJOMSC-UHFFFAOYSA-N
SMILES CC(=O)C1=CC2=C(C=C1N)OCO2
IUPAC Name 1-(6-amino-1,3-benzodioxol-5-yl)ethanone
Molecular Weight (g/mol) 179.175
PubChem CID 120000
Formula Weight 179.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6′-Amino-3′,4′-(methylenedioxy)acetophenone
Show More Show Less