missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2-naphthalenesulfonic Acid Monohydrate 98.0+%, TCI America™

Catalog No. A034825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A034825G 25 g
A0348500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A034825G Supplier TCI America Supplier No. A034825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-00-5
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD04038015
InChI Key SEMRCUIXRUXGJX-UHFFFAOYSA-N
PubChem CID 7116
ChEBI CHEBI:20700
IUPAC Name 6-aminonaphthalene-2-sulfonic acid
SMILES C1=CC2=C(C=CC(=C2)S(=O)(=O)O)C=C1N

Specifications

Quantity 25 g
Synonym Bronner′Cs Acid, 2-Naphthylamine-6-sulfonic Acid
Formula Weight 223.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2-naphthalenesulfonic Acid Monohydrate
EINECSNumber (4)-0493
TSCA Yes