missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2,4-lutidine 98.0+%, TCI America™

Catalog No. A07315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A07315G 5 g
A073125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A07315G Supplier TCI America Supplier No. A07315G
Only null left

Chemical Identifiers

CAS 5407-87-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00006322
InChI Key BRBUBVKGJRPRRD-UHFFFAOYSA-N
Synonym 2-amino-4,6-dimethylpyridine, 6-amino-2,4-lutidine, 2-pyridinamine, 4,6-dimethyl, 4,6-dimethyl-2-pyridinamine, 4,6-dimethyl-2-pyridylamine, pyridine, 2-amino-4,6-dimethyl, 2,4-lutidine, 6-amino, 2-amino-4,6-dimehtylpyridine, 4,6-dimethyl-2-aminopyridine, 2-amino-4,6-lutidine
PubChem CID 21507
IUPAC Name 4,6-dimethylpyridin-2-amine
SMILES CC1=CC(=NC(=C1)N)C

Specifications

Melting Point 67°C
Color White-Yellow
Boiling Point 231°C
Quantity 5 g
Formula Weight 122.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2,4-lutidine
RTECSNumber US1818000
TSCA Yes