missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-2,4-dimethoxypyrimidine 98.0+%, TCI America™

Catalog No. A22645G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22645G Supplier TCI America Supplier No. A22645G

Chemical Identifiers

CAS 3289-50-7
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.157
MDL Number MFCD00082802
InChI Key LNTJJKHTAZFVJJ-UHFFFAOYSA-N
Synonym 4-amino-2,6-dimethoxypyrimidine, 4-pyrimidinamine, 2,6-dimethoxy, 6-amino-2,4-dimethoxypyrimidine, 2,6-dimethoxy-4-aminopyrimidine, 2,6-dimethoxy-pyrimidin-4-ylamine, 2,4-dimethoxy-6-aminopyrimidine, 2,6-dimethoxy-4-pyrimidinamine, pyrimidine, 4-amino-2,6-dimethoxy, 2,6-dimethoxypyrimidine-4-ylamine, 2,6-dimethoxypyrimidin-4-amin
PubChem CID 76789
IUPAC Name 2,6-dimethoxypyrimidin-4-amine
SMILES COC1=NC(=NC(=C1)N)OC

Specifications

Melting Point 151°C
Quantity 5 g
Formula Weight 155.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Amino-2,4-dimethoxypyrimidine
EINECSNumber (5)-0931&(9)-0662&(9)-0104
TSCA Yes