missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-hexanol, 94%

Catalog No. AC294600500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294600500 50 g Glass bottle
AC294600100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294600500 Supplier Thermo Scientific Chemicals Supplier No. 294600500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4048-33-3 | C6H15NO | 117.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4048-33-3
Molecular Formula C6H15NO
Molecular Weight (g/mol) 117.19
MDL Number MFCD00008241
InChI Key SUTWPJHCRAITLU-UHFFFAOYSA-N
Synonym 6-amino-1-hexanol, 1-hexanol, 6-amino, 6-aminohexanol, 1-amino-6-hexanol, hexanol, 6-amino, 6-hydroxyhexylamine, aminohexyl alcohol, amidohexylalkohol, amidohexylalkohol german, 6-amino-hexan-1-ol
PubChem CID 19960
IUPAC Name 6-aminohexan-1-ol
SMILES C(CCCO)CCN

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 53°C to 59°C
Color Yellow to Brown
Boiling Point 233.0°C
Flash Point 133°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula H2N(CH2)6OH
Quantity 50 g
Beilstein 04, II, 748
Solubility Information Solubility in water: soluble
Formula Weight 117.19
Percent Purity 94%
Physical Form Crystals
Chemical Name or Material 6-Amino-1-hexanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
EINECSNumber 223-748-1

RUO – Research Use Only