missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-2H-1,4-benzoxazin-3(4H)-one 98.0+%, TCI America™

Catalog No. A27555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27555G 5 g
A275525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27555G Supplier TCI America Supplier No. A27555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 26518-71-8
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.186
MDL Number MFCD01321313
InChI Key BKJFWHFUERNXLJ-UHFFFAOYSA-N
PubChem CID 3641956
IUPAC Name 6-acetyl-4H-1,4-benzoxazin-3-one
SMILES CC(=O)C1=CC2=C(C=C1)OCC(=O)N2

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 5 g
Synonym 6-acetyl-2h-1,4-benzoxazin-3 4h-one, 6-acetyl-2h-benzo b 1,4 oxazin-3 4h-one, 2h-1,4-benzoxazin-3 4h-one, 6-acetyl, 6-acetyl-2,4-dihydro-1,4-benzoxazin-3-one, acmc-1co8k, 6-acetyl-4h-benzo 1,4 oxazin-3-one, 6-acetyl-2h-1,4-benzoxazine-3 4h-one, 2h-1,4-benzoxazin-3 4h-one,6-acetyl, 6-acetyl-2h-1,4-benzoxazin-3 4 h-one
Formula Weight 191.19
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-Acetyl-2H-1,4-benzoxazin-3(4H)-one