missing translation for 'onlineSavingsMsg'
Learn More

6-Acetyl-1,4-benzodioxane 98.0+%, TCI America™

Numéro de catalogue. A16295G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
A16295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. A16295G Fournisseur TCI America Code fournisseur A16295G
Il en reste null

Identifiants chimiques

CAS 2879-20-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00006823
InChI Key HGVWMTAIIYNQSI-UHFFFAOYSA-N
Synonym 6-acetyl-1,4-benzodioxane, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone, 1,4-benzodioxan-6-yl methyl ketone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one, 6-acetyl-1,4-benzodioxan, ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl, ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl, 1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethan-1-one, 6-acetyl-2h,3h-benzo e 1,4-dioxin
PubChem CID 76143
IUPAC Name 1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-one
SMILES CC(=O)C1=CC=C2OCCOC2=C1

Spécifications

Melting Point 84°C
Color White-Yellow
Quantity 5 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Acetyl-1,4-benzodioxane