missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxyquinazoline-2,4-dione, 98%

Catalog No. p-7044833
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0841109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0841109 Supplier Thermo Scientific Chemicals Supplier No. L0841109

CAS: 28888-44-0 | C10H10N2O4 | 222.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28888-44-0
Molecular Formula C10H10N2O4
Molecular Weight (g/mol) 222.20
MDL Number MFCD00023889
InChI Key KWNQIIMVPSMYEM-UHFFFAOYSA-N
Synonym 6,7-dimethoxyquinazoline-2,4-dione, 2,4-dihydroxy-6,7-dimethoxyquinazoline, 6,7-dimethoxyquinazoline-2,4 1h,3h-dione, 6,7-dimethoxyquinazoline-2,4-diol, 6,7-dimethoxy-1,2,3,4-tetrahydroquinazoline-2,4-dione, 6,7-dimethoxy-1h,3h-2,4-quinazolindione, 2,4 1h,3h-quinazolinedione, 6,7-dimethoxy, 6,7-dimethoxy-2,4-quinazolinedione, 2,4-dihydroxy-6,7-dimethoxy quinazoline, 6,7-dimethoxy-2,4-dihydroxy quinazoline
PubChem CID 120081
IUPAC Name 6,7-dimethoxy-1H-quinazoline-2,4-dione
SMILES COC1=C(OC)C=C2C(=O)NC(=O)NC2=C1

Specifications

Melting Point >300°C
Quantity 10 g
Beilstein 204597
Formula Weight 222.2
Percent Purity 98%
Chemical Name or Material 6,7-Dimethoxyquinazoline-2,4-dione
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 249-288-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only