missing translation for 'onlineSavingsMsg'
Learn More

6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride, 97%

Catalog No. AAB2246922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2246922 100 g
AAB2246906 5 g
AAB2246914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2246922 Supplier Thermo Scientific Chemicals Supplier No. B2246922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2328-12-3 | C11H16ClNO2 | 229.70 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2328-12-3
Molecular Formula C11H16ClNO2
Molecular Weight (g/mol) 229.70
MDL Number MFCD00012744
InChI Key SHOWAGCIRTUYNA-UHFFFAOYSA-N

Specifications

Melting Point 256°C to 260°C
Quantity 100 g
Beilstein 3634126
Synonym citrate iron, fe citrate
SMILES [Cl-].COC1=CC2=C(C[NH2+]CC2)C=C1OC
IUPAC Name 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinolin-2-ium chloride
PubChem CID 22178220
Formula Weight 229.71
Percent Purity 97%
Chemical Name or Material 6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber NX5017980
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only