missing translation for 'onlineSavingsMsg'
Learn More

6,6-Diphenylfulvene 98.0+%, TCI America™

Catalog No. D27875G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27875G 5 g
D27871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27875G Supplier TCI America Supplier No. D27875G
Only null left

Chemical Identifiers

CAS 2175-90-8
Molecular Formula C18H14
Molecular Weight (g/mol) 230.31
MDL Number MFCD00001419
InChI Key BULLHRADHZGONG-UHFFFAOYSA-N
Synonym 5-(Diphenylmethylene)-1,3-cyclopentadiene
PubChem CID 101236
IUPAC Name [cyclopenta-2,4-dien-1-ylidene(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(=C2C=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 81°C
Quantity 5 g
Formula Weight 230.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,6-Diphenylfulvene