missing translation for 'onlineSavingsMsg'
Learn More

6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone 98.0+%, TCI America™

Catalog No. A29191G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29191G Supplier TCI America Supplier No. A29191G
Only null left

Chemical Identifiers

CAS 36725-28-7
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.245
MDL Number MFCD01692712
InChI Key GDMRFHZLKNYRRO-UHFFFAOYSA-N
PubChem CID 161928
IUPAC Name 3-(4-aminophenyl)-4-methyl-4,5-dihydro-1H-pyridazin-6-one
SMILES CC1CC(=O)NN=C1C2=CC=C(C=C2)N

Specifications

Melting Point 196°C
Quantity 1 g
Formula Weight 203.25
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 6-(4-Aminophenyl)-4,5-dihydro-5-methyl-3(2H)-pyridazinone