missing translation for 'onlineSavingsMsg'
Learn More

6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chroman, 97%, Thermo Scientific™

Catalog No. CC32839CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC32839CB 250mg Amber Glass Bottle
CC32839DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC32839CB Supplier Thermo Scientific Chemicals Supplier No. CC32839CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1002727-88-9
Molecular Formula C15H21BO3
Molecular Weight (g/mol) 260.14
MDL Number MFCD12028565
InChI Key NDSHAELUPJMEBM-UHFFFAOYSA-N
Synonym 2-chroman-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl chroman, 2-3,4-dihydro-2h-1-benzopyran-6-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, chroman-6-boronic acid, pinacol ester, amtb667, chroman-6-boronic acid pinacol ester, chroman-6-ylboronic acid pinacol ester, 3,4-dihydro-2h-chromene-6-boronic acid, pinacol ester, 3,4-dihydro-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2h-chromene
PubChem CID 43811034
IUPAC Name 2-(3,4-dihydro-2H-chromen-6-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(OCCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 260.14
Percent Purity 97%
Chemical Name or Material 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chroman
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.