missing translation for 'onlineSavingsMsg'
Learn More

6,13-Diphenylpentacene 95.0+%, TCI America™

Catalog No. D4469200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4469200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4469200MG Supplier TCI America Supplier No. D4469200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 76727-11-2
Molecular Formula C34H22
Molecular Weight (g/mol) 430.55
MDL Number MFCD22572658
InChI Key PFCSVQKCECNEAZ-UHFFFAOYSA-N
PubChem CID 11201250
IUPAC Name 6,13-diphenylpentacene
SMILES C1=CC=C(C=C1)C1=C2C=C3C=CC=CC3=CC2=C(C2=CC=CC=C2)C2=CC3=CC=CC=C3C=C12

Specifications

Melting Point 254°C
Color Black-Purple
Quantity 200 mg
Formula Weight 430.55
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6,13-Diphenylpentacene