Learn More
World Precision Cover slips, pkg. 100-502040
Supplier:  World Precision WP502040
Encompass
These cover slips (made of German glass) can be used for growing and culturing cells that normally have poor adhesion to plastic surfaces. They are small enough to be placed in the micro plate or other cell culture devices. The 5 mm size will fit inside the 96-well culture plate and leave enough room to pick it up from the bottom of the well with forceps. The 8mm size fits inside the 24-well plates.
Numéro de catalogue. NC0134776
Il en reste null