Learn More
Chemglass Inc Vacuum Trap, 500mL Capacity, Fits 1900mL Dewar Flask, 310mm OAH x 130mm Width, Complete
Numéro de catalogue. CG452901
Encompass
Vacuum Trap, 500mL Capacity, Fits 1900mL Dewar Flask, 310mm OAH x 130mm Width, Complete. Trap is designed for large volume solvent removal by vacuum. The 40/35 standard taper joint connects trap body with stopper. Connections are 1/2in O.D. Trap is designed to fit into standard CG-1593 through CG-1595 Dewar flasks.
Numéro de catalogue. CG452901 Fournisseur Chemglass Inc Code fournisseur CG452901
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Vacuum Trap, 500mL Capacity, Fits 1900mL Dewar Flask, 310mm OAH x 130mm Width, Complete. Trap is designed for large volume solvent removal by vacuum. The 40/35 standard taper joint connects trap body with stopper. Connections are 1/2in O.D. Trap is designed to fit into standard CG-1593 through CG-1595 Dewar flasks.