missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5000 Series 0.31μm to 0.93μm Polymer Particle Suspensions

Catalog No. 09982193
Change view
Click to view available options
Diameter (Metric):
0.31 μm
0.33 μm
0.36 μm
0.43 μm
0.49 μm
0.5 μm
0.51 μm
0.52 μm
0.6 μm
0.65 μm
0.67 μm
0.75 μm
0.82 μm
0.87 μm
0.93 μm
Quantity:
15 mL
100 mL
1000 mL
37 product options available for selection
Product selection table with 37 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Quantity
09982193 0.75 μm 100 mL
09980284 0.31 μm 15 mL
09980285 0.31 μm 100 mL
09980286 0.31 μm 1000 mL
09980287 0.33 μm 15 mL
09980288 0.33 μm 100 mL
09980289 0.33 μm 1000 mL
09980290 0.36 μm 15 mL
09980291 0.36 μm 100 mL
09980292 0.36 μm 1000 mL
09980293 0.43 μm 15 mL
09980294 0.43 μm 100 mL
09980295 0.43 μm 1000 mL
09980299 0.49 μm 15 mL
09980300 0.49 μm 100 mL
09980301 0.49 μm 1000 mL
09980302 0.5 μm 15 mL
09980303 0.5 μm 100 mL
09980304 0.5 μm 1000 mL
09980305 0.51 μm 15 mL
09980306 0.51 μm 100 mL
09980307 0.51 μm 1000 mL
09980308 0.52 μm 15 mL
09980309 0.52 μm 100 mL
09980311 0.6 μm 15 mL
09980312 0.6 μm 100 mL
09980313 0.6 μm 1000 mL
09980315 0.65 μm 100 mL
09980316 0.65 μm 1000 mL
09980317 0.67 μm 15 mL
09982192 0.75 μm 15 mL
09980323 0.82 μm 15 mL
09980324 0.82 μm 100 mL
09980327 0.87 μm 100 mL
09980328 0.87 μm 1000 mL
09980330 0.93 μm 100 mL
09980331 0.93 μm 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 37 options available.
37 missing translation for 'options'
Catalog No. 09982193 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' 5074B

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 5000 Series Polymer Particle Suspensions

Meet the need for particulate materials with a variety of particle sizes and properties with Thermo Scientific™ 5000 Series Polymer Particle Suspensions, of special interest for research in light scattering, microporous filter checking and aerosol particle generation.

Features:

  • Density of 1.05g/cm3 and a refractive index of 1.59 at 589nm
  • Diameters measured by optical microscopy, photon correlation spectroscopy or light scattering
  • Packaged as aqueous suspensions at 10% solids by weight
  • Range up to 3.1 microns in size
  • For larger particles, refer to our 7000 Series Copolymer Particle Suspensions

Ordering Information:

  • 15mL, 100mL, and 1000mL bottle sizes available
  • 15mL (“A” Part No. bottles), 100mL (B bottles) and 1000mL (C bottles) are packaged to order; example (Part No. 5003B and 5003C).
  • Please allow 10 working days when ordering these fill volumes.

Includes:

Material Safety Data Sheet (MSDS) and general product handling insert sheet

Recommended for:

High concentration polymer particles < 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications.

Specifications

Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles useful for such applications as filter evaluation, checking and testing, light scattering research, fluid mechanics research, aerosol particle generation, dispersion studies, and many other research and development projects. They are not intended for use in instrument calibration.
Color White or Colorless
For Use With (Application) Aerosol Particle Generation, Dispersion, Filter Evaluation, Light Scattering, Particle Generation
Material Polystyrene
Product Type Polymer Particle Suspensions
Includes General product handling insert sheet
Packaging 100 mL
Particle Density 1.05g/cm3
Quantity 100 mL
Refractive Index 1.59 at 589nm (25°C)
Stability Product stable for a minimum of 24 months
Diameter (Metric) 0.75 μm
Show More Show Less