Learn More
Fisherbrand™ 50 Well Hinged Tube Storage Box
Catalog No. 14127169
Change view
Click to view available options
Color:
Assorted (Blue, Green, Orange, Purple, Yellow)
Natural
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14127169 Assorted (Blue, Green, Orange, Purple, Yellow)
14127118 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14127169 Supplier Fisherbrand™ Supplier No. 120033F2
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
A polypropylene tube storage box designed for 1.5 mL and 2 mL tubes and cryogenic vials
  • The 50-Well Microtube Storage Box is ideal for sample or case study storage
  • Made from durable polypropylene, suitable for regular lab use.
  • Uniform wells with alphanumeric coordinates help you locate samples quickly tube capacity is shown on the outside.
  • Frosted region included for easy writing or label placement.
  • Available in assorted vibrant translucent colors or natural versions depending on supplier.
  • Microtube storage boxes
  • Hold 1.5 and 2.0 mL microcentrifuge tubes and cryogenic vials
  • Made of translucent, vibrantly colored polypropylene allows for visibility inside box and color coding identification
  • Tube capacity is indicated on the outside the box within a distinctive wave design
  • Quick visual identification with uniform wells and alphanumeric location identification along with a frosted labeling area
  • Easy open and secure lid fitted with either friction fit or snap clasp designs
  • Stackable, saving valuable space in the lab
Specifications
Closure Type Hinged Lid
Color Assorted (Blue, Green, Orange, Purple, Yellow)
Material Polypropylene
Shape Rectangular
Type Cryo Storage Box
Holds 50 Tubes and Vials
Quantity 5 Pk.
Sterility Non-sterile
Tube Array 5 x 10
Withstand Temperature (Metric) -80°C
Dimensions (L x W x H) 5.6 x 3.6 x 2.2 in.
Show More Show Less