missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)nicotinic acid, 97%, Thermo Scientific Chemicals

Catalog No. AAH6390603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6390603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6390603 Supplier Thermo Scientific Chemicals Supplier No. H6390603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 131747-40-5 | Molecular Formula: C7H4F3NO2 | Molecular Weight: 191.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 131747-40-5
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.11
MDL Number MFCD07774128
InChI Key RKFDCELCLIZRRH-UHFFFAOYSA-N
Synonym 5-trifluoromethyl nicotinic acid, 5-trifluoromethyl pyridine-3-carboxylic acid, 5-trifluoromethylinicotinic acid, 5-trifluoromethyl-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl-nicotinic acid, 3-pyridinecarboxylicacid, 5-trifluoromethyl, 5-trifluoromethy pyridine-3-carboxylic acid
PubChem CID 10976305
IUPAC Name 5-(trifluoromethyl)pyridine-3-carboxylic acid
SMILES OC(=O)C1=CC(=CN=C1)C(F)(F)F

Specifications

Melting Point 184°C to 189°C
Color White
Quantity 1 g
UN Number UN2811
Formula Weight 191.11
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-(Trifluoromethyl)nicotinic acid
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only