missing translation for 'onlineSavingsMsg'
Learn More

5-(Trifluoromethyl)-2-pyridinecarboxylic Acid 98.0+%, TCI America™

Catalog No. T29755G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29755G 5 g
T29751G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T29755G Supplier TCI America Supplier No. T29755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 80194-69-0
Molecular Formula C7H4F3NO2
Molecular Weight (g/mol) 191.109
MDL Number MFCD04113632
InChI Key NJHGVAYLDHROPT-UHFFFAOYSA-N
Synonym 5-trifluoromethyl pyridine-2-carboxylic acid, 5-trifluoromethyl picolinic acid, 5-trifluoromethyl-2-pyridinecarboxylic acid, 5-trifluoromethyl-pyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 5-trifluoromethyl, 5-trifluoromethyl pyridine-2-carboxylicacid, pubchem15164, acmc-209pj2
PubChem CID 10910349
IUPAC Name 5-(trifluoromethyl)pyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1C(F)(F)F)C(=O)O

Specifications

Melting Point 137°C
Color White-Yellow
Quantity 5 g
Formula Weight 191.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(Trifluoromethyl)-2-pyridinecarboxylic Acid