missing translation for 'onlineSavingsMsg'
Learn More

5-tert-Butyl-2-methoxybenzeneboronic acid, 98+%, Thermo Scientific Chemicals

Catalog No. AAL2041503
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2041503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL2041503 Supplier Thermo Scientific Chemicals Supplier No. L2041503
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128733-85-7 | Molecular Formula: C11H17BO3 | Molecular Weight: 208.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128733-85-7
Molecular Formula C11H17BO3
Molecular Weight (g/mol) 208.064
MDL Number MFCD06201034
InChI Key LXFOJKCQSAYVMF-UHFFFAOYSA-N
Synonym 5-tert-butyl-2-methoxyphenyl boronic acid, 5-t-butyl-2-methoxyphenylboronic acid, 5-tert-butyl-2-methoxybenzeneboronic acid, 2-borono-4-tert-butyl anisole, acmc-1c4zr, 2-methoxy-5-tert-butylphenylboronic acid, 5-tert-butyl-2-methoxyphenyl boronicacid, boronic acid, b-5-1,1-dimethylethyl-2-methoxyphenyl
PubChem CID 14740579
IUPAC Name (5-tert-butyl-2-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)C(C)(C)C)OC)(O)O

Specifications

Melting Point 103°C to 106°C
Quantity 1 g
Beilstein 4181890
Formula Weight 208.07
Percent Purity ≥98%
Chemical Name or Material 5-tert-Butyl-2-methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only