missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Sulfosalicylic Acid Dihydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific FLA297100
Specifications
| Sulfosalicylic Acid Dihydrate | |
| 5965-83-3 | |
| White | |
| 0.1% max. | |
| Glass Bottle | |
| 1-(OH)-2-(COOH)-4-SO3H-C6H3·2H2O | |
| 100 g | |
| 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd | |
| NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
| 2-hydroxy-5-sulfobenzoic acid; bis(methane) | |
| 2723734 | |
| 99.0 to 101.0% | |
| Powder or Solid |
| 5-Sulfosalicylic Acid Dihydrate | |
| 106°C | |
| 0.5 | |
| 99.0 to 101.0% | |
| C9H14O6S | |
| MFCD00007508,MFCD00149540 | |
| 2585 | |
| 0.02% max. Insoluble Matter | |
| C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O | |
| 250.27 | |
| 254.22 | |
| Certified ACS |
Chemical Identifiers
| 5965-83-3 | |
| 250.27 | |
| NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
| 2723734 | |
| C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O |
| C9H14O6S | |
| MFCD00007508,MFCD00149540 | |
| 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd | |
| 2-hydroxy-5-sulfobenzoic acid; bis(methane) |
Safety and Handling
DOTInformation : DOT Class 8, : Corrosive