missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid Dihydrate (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-193672
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A297500 500 g Glass Bottle
A2973 3 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A297500 Supplier Thermo Fisher Scientific Supplier No. FLA297500
Only null left

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
MDL Number MFCD00007508,MFCD00149540
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
PubChem CID 2723734
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

Alpha Vector Sulfosalicylic Acid Dihydrate
Melting Point 106°C
Color White
pH 0.5
Ignition Residue 0.1% max.
Assay Percent Range 99.0 to 101.0%
Packaging Glass Bottle
Linear Formula 1-(OH)-2-(COOH)-4-SO3H-C6H3·2H2O
Quantity 500 g
UN Number 2585
Solubility Information 0.02% max. Insoluble Matter
Formula Weight 254.22
Percent Purity 99.0 to 101.0%
Grade Certified ACS
Physical Form Powder or Solid
Chemical Name or Material 5-Sulfosalicylic Acid Dihydrate
Show More Show Less
DOTInformation DOT Class 8, : Corrosive