missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-SulfoSalicylic Acid Dihydrate, ACS Reagent, Reagents
5-SulfoSalicylic Acid Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C13354003A5
Specifications
| 5-SulfoSalicylic Acid Dihydrate, ACS Reagent | |
| White | |
| C9H14O6S | |
| 0.8 | |
| NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
| 2-hydroxy-5-sulfobenzoic acid; bis(methane) | |
| ACS |
| 5965-83-3 | |
| Poly Bottle | |
| 3 kg | |
| 2-Hydroxy-5-sulfobenzoic Acid dihydrate, Sulfosalicylic Acid dihydrate | |
| C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O | |
| 250.27 | |
| Solid |
Chemical Identifiers
| 5965-83-3 | |
| 250.27 | |
| 2-Hydroxy-5-sulfobenzoic Acid dihydrate, Sulfosalicylic Acid dihydrate | |
| C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O |
| C9H14O6S | |
| NFYHZVWMQHQKRU-UHFFFAOYSA-N | |
| 2-hydroxy-5-sulfobenzoic acid; bis(methane) |