missing translation for 'onlineSavingsMsg'
Learn More

5-SulfoSalicylic Acid Dihydrate, ACS Reagent, Reagents

Catalog No. p-200021011
Change view
Click to view available options
Quantity:
125 g
500 g
3 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337651 10 kg
01337650 125 g
01337649 3 kg
01337648 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01337651 Supplier Labchem Inc Supplier No. C133540010KG
Only null left

5-SulfoSalicylic Acid Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 5965-83-3
Molecular Formula C9H14O6S
Molecular Weight (g/mol) 250.27
InChI Key NFYHZVWMQHQKRU-UHFFFAOYSA-N
Synonym 2-Hydroxy-5-sulfobenzoic Acid dihydrate, Sulfosalicylic Acid dihydrate
IUPAC Name 2-hydroxy-5-sulfobenzoic acid; bis(methane)
SMILES C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O

Specifications

Color White
Packaging Poly Pail
Quantity 10 kg
Specific Gravity 0.8
Grade ACS
Physical Form Solid
Chemical Name or Material 5-SulfoSalicylic Acid Dihydrate, ACS Reagent