missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Ring NOGE (Novolac glycidyl ether) Mixture of Isomers, chain-like or branched, ≥90% (HPLC), MilliporeSigma™ Supelco™
Description
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
Specifications
Specifications
| Physical Form | Neat |
| Synonym | NOGE 5-Ring (Novolac glycidyl ether) mixture of isomers; Novolac glycidyl ether 5-Ring |
| Type | Analytical Standard |
| Quantity | 50 mg |
| Percent Purity | ≥90% (HPLC) |
| Grade | Analytical Standard |