missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinylboronic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC07412EA

Quantity 10g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC07412EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

109299-78-7
123.91
HZFPPBMKGYINDF-UHFFFAOYSA-N
2795193
OB(O)C1=CN=CN=C1
C4H5BN2O2
MFCD03002366,MFCD16661236
5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
(pyrimidin-5-yl)boronic acid

Specifications

5-Pyrimidinylboronic acid
97%
C4H5BN2O2
10g
HZFPPBMKGYINDF-UHFFFAOYSA-N
(pyrimidin-5-yl)boronic acid
2795193
97%
109299-78-7
Amber Glass Bottle
MFCD03002366,MFCD16661236
5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid
OB(O)C1=CN=CN=C1
123.91
123.91
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Warning

SDS
Documents

Documents