missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrimidinecarboxylic acid, 97%, Thermo Scientific™

Catalog No. CC07401DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC07401DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC07401DA Supplier Thermo Scientific Chemicals Supplier No. CC07401DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4595-61-3
Molecular Formula C5H4N2O2
Molecular Weight (g/mol) 124.10
MDL Number MFCD00856162
InChI Key IIVUJUOJERNGQX-UHFFFAOYSA-N
Synonym 5-pyrimidinecarboxylic acid, 5-carboxypyrimidine, pyrimidine-5-carboxilic acid, 5-pyrimidine carboxylic acid, pyrimidine-5-carboxylate acid, pyrimidine-5-carboxylicacid, pubchem2056, acmc-209k5a, 5-pyrimidyl carboxylic acid, pyrimidin-5-carboxylic acid
PubChem CID 78346
IUPAC Name pyrimidine-5-carboxylic acid
SMILES OC(=O)C1=CN=CN=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 124.1
Percent Purity 97%
Chemical Name or Material 5-Pyrimidinecarboxylic acid