missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyridin-2-ylthiophene-2-carbaldehyde, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Quantity | 250mg |
| Formula Weight | 189.24 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Pyridin-2-ylthiophene-2-carbaldehyde |