missing translation for 'onlineSavingsMsg'
Learn More

5-Pyrazolone 98.0+%, TCI America™

Catalog No. p-7135601
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2563200MG 200 mg
P25631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P2563200MG Supplier TCI America Supplier No. P2563200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 137-44-0
Molecular Formula C3H4N2O
Molecular Weight (g/mol) 84.08
MDL Number MFCD00127223
InChI Key ZRHUHDUEXWHZMA-UHFFFAOYSA-N
Synonym 2,4-Dihydro-3H-pyrazol-3-one, 1H-Pyrazol-5(4H)-one
PubChem CID 316289
IUPAC Name 4,5-dihydro-1H-pyrazol-5-one
SMILES O=C1CC=NN1

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 200 mg
Formula Weight 84.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Pyrazolone