missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyltetrazole 99.0+%, TCI America™

Catalog No. P1109500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1109500G 500 g
P110925G 25 g
P1109100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P1109500G Supplier TCI America Supplier No. P1109500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 18039-42-4
Molecular Formula C7H6N4
Molecular Weight (g/mol) 146.153
MDL Number MFCD00022388
InChI Key MARUHZGHZWCEQU-UHFFFAOYSA-N
Synonym 5-phenyl-1h-tetrazole, 5-phenyltetrazole, 1h-tetrazole, 5-phenyl, 2h-tetrazole, 5-phenyl, phenyltetrazole, 5-phenyl tetrazole, expandex 5pt, expandex ox 5pt, kempore 50xpt, 5-phenyltetrazole van
PubChem CID 87425
IUPAC Name 5-phenyl-2H-tetrazole
SMILES C1=CC=C(C=C1)C2=NNN=N2

Specifications

Melting Point 215°C
Color White
Quantity 500 g
UN Number 1325
Formula Weight 146.15
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenyltetrazole
EINECSNumber (5)-6042
RTECSNumber XF7538750
TSCA Yes