missing translation for 'onlineSavingsMsg'
Learn More
Learn More
acide 5-phénylpyridine-2-carboxylique, 95%, Thermo Scientific™
Spécifications
Spécifications
| CAS Min % | 94.5 |
| CAS Max % | 100.0 |
| Assay Percent Range | 95% |
| Packaging | Bouteille en verre ambrée |
| Quantity | 1 g |
| Formula Weight | 199.21 |
| Percent Purity | 95% |
| Chemical Name or Material | 5-Phenylpyridine-2-carboxylic acid |
Sécurité et manipulation
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement: Harmful if swallowed. Causes skin irritation. May cause respiratory irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |