missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylisoxazole-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. P20205G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20205G 5 g
P20201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20205G Supplier TCI America Supplier No. P20205G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 14441-90-8
Molecular Formula C10H7NO3
Molecular Weight (g/mol) 189.17
MDL Number MFCD00464221
InChI Key XJYOBHXWBRKOQO-UHFFFAOYSA-N
Synonym 5-phenylisoxazole-3-carboxylic acid, 5-phenyl-isoxazole-3-carboxylic acid, 5-phenyl-3-isoxazolecarboxylic acid, 3-isoxazolecarboxylic acid, 5-phenyl, 3-isoxazolecarboxylicacid, 5-phenyl, 5-phenylisoxazole-3-carboxylicacid, 5-phenylisoxazol-3-carboxylic acid
PubChem CID 151916
IUPAC Name 5-phenyl-1,2-oxazole-3-carboxylic acid
SMILES C1=CC=C(C=C1)C2=CC(=NO2)C(=O)O

Specifications

Melting Point 160°C
Quantity 5 g
Formula Weight 189.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenylisoxazole-3-carboxylic Acid