missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylcyclohexane-1,3-dione, 97%

Numéro de catalogue. AAA1213614
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAA1213614 25 g
AAA1213606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAA1213614 Fournisseur Thermo Scientific Chemicals Code fournisseur A1213614
Il en reste null

CAS: 493-72-1 | C12H12O2 | 188.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 493-72-1
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.23
MDL Number MFCD00051846
InChI Key ZFVDSCASCRHXCP-SNVBAGLBSA-N
PubChem CID 568962
IUPAC Name 5-phenylcyclohexane-1,3-dione
SMILES OC1=CC(=O)C[C@@H](C1)C1=CC=CC=C1

Spécifications

Melting Point ∼185°C (decomposition)
Quantity 25 g
Beilstein 1103470
Synonym 5-phenyl-1,3-cyclohexanedione, 5-phenyl-cyclohexane-1,3-dione, 1,3-cyclohexanedione, 5-phenyl, acmc-1anml, 5-phenylcyclohexan-1,3-dione, 1,3-dioxo-5-phenylcyclohexane, 5phenylcyclohexane-1, 3-dione, 1,3-cyclohexanedione,5-phenyl, 5-phenyl-cyclohexane-1, 3-dione
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material 5-Phenylcyclohexane-1,3-dione
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only