missing translation for 'onlineSavingsMsg'
Learn More

5-(p-Tolyl)-1H-tetrazole 98.0+%, TCI America™

Catalog No. T24865G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24865G 5 g
T248625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24865G Supplier TCI America Supplier No. T24865G
Only null left

Chemical Identifiers

CAS 24994-04-5
Molecular Formula C8H8N4
Molecular Weight (g/mol) 160.18
MDL Number MFCD01318164
InChI Key BCCJIAZPYBJASR-UHFFFAOYSA-N
Synonym 5-4-methylphenyl-1h-tetrazole, 5-p-tolyl-1h-tetrazole, 5-4-methylphenyl-2h-tetrazole, 5-4-methylphenyl-2h-1,2,3,4-tetrazole, 5-p-tolyl-2h-tetrazole, 5-4-methylphenyl-2h-1,2,3,4-tetraazole, 5-p-methylphenyl-1h-tetrazole, 5-4-methylphenyl tetrazole
PubChem CID 285164
IUPAC Name 5-(4-methylphenyl)-2H-tetrazole
SMILES CC1=CC=C(C=C1)C2=NNN=N2

Specifications

Melting Point 248°C
Color White-Yellow
Quantity 5 g
Formula Weight 160.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(p-Tolyl)-1H-tetrazole