missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-methylamine (mixture of isomers) 98.0+%, TCI America™

Catalog No. N09075G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09075G 5 g
N090725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09075G Supplier TCI America Supplier No. N09075G

Chemical Identifiers

CAS 95-10-3
Molecular Formula C8H13N
Molecular Weight (g/mol) 123.199
MDL Number MFCD07369043
InChI Key XLBALIGLOMYEKN-UHFFFAOYSA-N
Synonym 5-(Aminomethyl)bicyclo[2.2.1]hept-2-ene, 5-(Aminomethyl)norbornene, 5-Norbornene-2-methanamine
PubChem CID 247182
IUPAC Name 5-bicyclo[2.2.1]hept-2-enylmethanamine
SMILES C1C2CC(C1C=C2)CN

Specifications

Color Yellow
Quantity 5 g
UN Number 2735
Formula Weight 123.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Norbornene-2-methylamine (mixture of isomers)