missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornene-2-carboxamide (mixture of isomers) 98.0+%, TCI America™

Catalog No. N09115G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N09115G 5 g
N091125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N09115G Supplier TCI America Supplier No. N09115G
Only null left

Chemical Identifiers

CAS 95-17-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00187896
InChI Key ZTUUVDYQBLRAAC-UHFFFAOYSA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2-carboxamide
PubChem CID 96112
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carboxamide
SMILES C1C2CC(C1C=C2)C(=O)N

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2-carboxamide (mixture of isomers)