missing translation for 'onlineSavingsMsg'
Learn More

5-Norbornen-2-ol, mixture of endo and exo, 98+%

Catalog No. AAH5699414
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5699414 25 g
AAH5699403 1 g
AAH5699406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5699414 Supplier Thermo Scientific Chemicals Supplier No. H5699414
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 13080-90-5 | C7H10O | 110.156 g/mol

It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in High Performance Liquid Chromatography. (HPLC) and Nuclear Magnetic Resonance (NMR).

Solubility
Slightly soluble in water

Notes
Store at 2-8°C. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 13080-90-5
Molecular Formula C7H10O
Molecular Weight (g/mol) 110.156
MDL Number MFCD00167566
InChI Key MKOSBHNWXFSHSW-UHFFFAOYSA-N
Synonym 5-norbornene-2-ol, bicyclo 2.2.1 hept-5-en-2-ol, 5-norbornen-2-ol, 5-norbornen-2-ol, exo, exo-2-norbornenol, bicyclo 2.2.1 hept-2-en-5-ol, bicyclo 2.2.1 hept-5-en-2-ol, exo, 5-norbornen-2-ol exo
PubChem CID 96066
IUPAC Name bicyclo[2.2.1]hept-2-en-5-ol
SMILES C1C2CC(C1C=C2)O

Specifications

Melting Point 104°C to 106°C
Flash Point 51°C (124°F)
Quantity 25 g
UN Number UN1325
Solubility Information Slightly soluble in water
Formula Weight 110.15
Percent Purity 99%
Chemical Name or Material 5-Norbornen-2-ol, mixture of endo and exo
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber 235-987-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only