missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furoic acid, 98+%

Catalog No. AAA1059722
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1059722 100 g
AAA1059706 5 g
AAA1059714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1059722 Supplier Thermo Scientific Chemicals Supplier No. A1059722
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 645-12-5 | C5H3NO5 | 157.081 g/mol

5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-furoic acid was used in the preparation of [1,2,4]triazolo[3,4-b][1,3,4]thiadiazines and [1,2,4]triazolo[3,4-b][1,3,4] thiadiazoles. The FT-IR and FT-Raman spectra of 5-Nitro-2-furoic acid has also been studied.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 645-12-5
Molecular Formula C5H3NO5
Molecular Weight (g/mol) 157.081
MDL Number MFCD00003240
InChI Key IODMEDPPCXSFLD-UHFFFAOYSA-N
Synonym 5-nitro-2-furoic acid, nitrofurate, 5-nitrofuroic acid, 2-furoic acid, 5-nitro, 5-nitro-2-furancarboxylic acid, 5-nitropyromucate, 5-nitrofurancarboxylic acid, 2-furancarboxylic acid, 5-nitro, ccris 1197, 5-nitropyromucic acid
PubChem CID 12577
IUPAC Name 5-nitrofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)[N+](=O)[O-])C(=O)O

Specifications

Melting Point 186°C to 188°C
Quantity 100 g
Beilstein 139373
Solubility Information Slightly soluble in water.
Formula Weight 157.08
Percent Purity ≥98%
Chemical Name or Material 5-Nitro-2-furoic acid
Hazard Category H315-H319-H335-H341
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
EINECSNumber 211-432-6
RTECSNumber LV2012000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only