missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-2-furaldehyde, 99%

Catalog No. AAA1061818
Change view
Click to view available options
Quantity:
25 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1061818 50 g
AAA1061814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1061818 Supplier Thermo Scientific Chemicals Supplier No. A1061818
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 698-63-5 | C5H3NO4 | 141.082 g/mol

5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.

Chemical Identifiers

CAS 698-63-5
Molecular Formula C5H3NO4
MDL Number MFCD00003230
InChI Key SXINBFXPADXIEY-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde, 5-nitrofurfural, nitrofurfural, 2-furancarboxaldehyde, 5-nitro, 5-nitrofurfuraldehyde, 5-nitro-2-furfural, 2-furaldehyde, 5-nitro, 5-nitrofuraldehyde, 5-nitro-2-furancarboxaldehyde, furfural, 5-nitro
PubChem CID 12774
ChEBI CHEBI:34459
IUPAC Name 5-nitrofuran-2-carbaldehyde

Specifications

Melting Point 34°C to 38°C
Density 1.349
Boiling Point 228°C
Flash Point 33°C (91°F)
Refractive Index 1.59
Quantity 50 g
Beilstein 120539
Sensitivity Air and light sensitive
Solubility Information Sparingly soluble in water.
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=O
Molecular Weight (g/mol) 141.082
Formula Weight 141.09
Percent Purity 99%
Chemical Name or Material 5-Nitro-2-furaldehyde
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 211-816-3
RTECSNumber LT7200000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only