missing translation for 'onlineSavingsMsg'
Learn More

5-Methyluridine 98.0+%, TCI America™

Catalog No. M14051G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14051G 1 g
M14055G 5 g
M140525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M14051G Supplier TCI America Supplier No. M14051G

Chemical Identifiers

CAS 1463-10-1
Molecular Formula C10H14N2O6
Molecular Weight (g/mol) 258.23
MDL Number MFCD00006535
InChI Key DWRXFEITVBNRMK-JXOAFFINSA-N
Synonym 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine
PubChem CID 445408
ChEBI CHEBI:45996
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 183°C
Color White
Quantity 1 g
Formula Weight 258.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyluridine
EINECSNumber (5)-6843
TSCA No