missing translation for 'onlineSavingsMsg'
Learn More

5-Methyluridine, 99%

Catalog No. AAH6425922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6425922 100 g
AAH6425914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6425922 Supplier Thermo Scientific Chemicals Supplier No. H6425922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1463-10-1 | C10H14N2O6 | 258.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1463-10-1
Molecular Formula C10H14N2O6
Molecular Weight (g/mol) 258.23
MDL Number MFCD00006535
InChI Key DWRXFEITVBNRMK-JXOAFFINSA-N
Synonym 5-methyluridine, ribothymidine, thymine riboside, ribosylthymine, uridine, 5-methyl, ribosylthymidine, 5-methyl-uridine, beta-d-ribofuranoside, thymine ribofuranoside, 1-beta-d-ribofuranosyl thymine
PubChem CID 445408
ChEBI CHEBI:45996
IUPAC Name 1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methylpyrimidine-2,4-dione
SMILES CC1=CN(C(=O)NC1=O)C2C(C(C(O2)CO)O)O

Specifications

Melting Point 183°C to 184°C
Color White
Quantity 100 g
Beilstein 90164
Formula Weight 258.23
Percent Purity 99%
Physical Form Solid
Chemical Name or Material 5-Methyluridine
EINECSNumber 215-973-9
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only