missing translation for 'onlineSavingsMsg'
Learn More

5-Methylumbelliferone 95.0+%, TCI America™

Catalog No. M29481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M29481G Supplier TCI America Supplier No. M29481G
Only null left

Chemical Identifiers

CAS 7249-26-5
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
MDL Number MFCD00488356
InChI Key QNDMQTVAGWUUDS-UHFFFAOYSA-N
Synonym 7-Hydroxy-5-methylcoumarin
PubChem CID 5355639
IUPAC Name 7-hydroxy-5-methylchromen-2-one
SMILES CC1=CC(=CC2=C1C=CC(=O)O2)O

Specifications

Melting Point 248°C
Quantity 1 g
Formula Weight 176.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylumbelliferone