missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-sulfonyl chloride, 97%, Thermo Scientific™

Numéro de catalogue. CC65003DA
Modifier l'affichage
Click to view available options
Quantité:
1g
5g
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
CC65003DA 1g
CC65003DE 5g
CC65003EA 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. CC65003DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC65003DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 55854-45-0
Formule moléculaire C5H5ClO2S2
Poids moléculaire (g/mol) 196.66
Numéro MDL MFCD05864499
Clé InChI QGGRRHYGHGJEKP-UHFFFAOYSA-N
Synonyme 5-methyl-2-thiophenesulfonyl chloride, 2-thiophenesulfonyl chloride, 5-methyl, 5-methylthiophene-2-sulfonylchloride, 5-methylthiophene-2-sulphonyl chloride, 5-methyl-2-thienylsulfonyl chloride, chloro 5-methyl 2-thienyl sulfone, acmc-209lpu, 5-methylthiophenesulphonyl chloride, 5-methyl-thiophenesulfonyl chloride, 2-chlorosulphonyl-5-methylthiophene
PubChem CID 12400112
Nom de l’IUPAC 5-methylthiophene-2-sulfonyl chloride
SOURIRES CC1=CC=C(S1)S(Cl)(=O)=O

Spécifications

Nom ou substance chimique 5-Methylthiophene-2-sulfonyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 95%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 196.68
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.