missing translation for 'onlineSavingsMsg'
Learn More

5-Methylthiophene-2-carbonyl chloride, 97%, Thermo Scientific™

Numéro de catalogue. CC65002DA
Modifier l'affichage
Click to view available options
Quantité:
250mg
1g
Emballage:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité Emballage
CC65002DA 1g Amber Glass Bottle
CC65002CB 250mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. CC65002DA Fournisseur Thermo Scientific Chemicals Code fournisseur CC65002DA

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

Identifiants chimiques

CAS 31555-59-6
Formule moléculaire C6H5ClOS
Poids moléculaire (g/mol) 160.615
Numéro MDL MFCD00130092
Clé InChI DJCGIWVUKCNCAT-UHFFFAOYSA-N
Synonyme 5-methyl-thiophene-2-carbonyl chloride, 5-methyl-2-thiophenecarbonyl chloride, 2-thiophenecarbonyl chloride, 5-methyl, 2-thiophenecarbonylchloride, 5-methyl, 5-methyl-2-thenoyl chloride, 2-chlorocarbonyl-5-methylthiophene, 5-methyl-thiophene-carboxylicacid chloride, 5-methylthiophene-2-carboxylic acid chloride, 5-methyl-thiophene-2-carboxylic acid chloride
PubChem CID 2760083
Nom de l’IUPAC 5-methylthiophene-2-carbonyl chloride
SOURIRES CC1=CC=C(S1)C(=O)Cl

Spécifications

Nom ou substance chimique 5-Methylthiophene-2-carbonyl chloride
CAS % minimum 96.5
CAS % maximal 100.0
Fourchette de pourcentages d’essai 97%
Emballage Amber Glass Bottle
Quantité 1g
Poids de formule 160.62
Pourcentage de pureté 97%
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.