missing translation for 'onlineSavingsMsg'
Learn More

5-(Methylthio)thiophene-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. M24481G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24481G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24481G Supplier TCI America Supplier No. M24481G
Only null left

Chemical Identifiers

CAS 24445-35-0
Molecular Formula C6H6OS2
Molecular Weight (g/mol) 158.233
MDL Number MFCD00173744
InChI Key RKXYTFTZODXDEF-UHFFFAOYSA-N
Synonym 5-methylthio thiophene-2-carbaldehyde, 5-methylthio thiophene-2-carboxaldehyde, 5-methylsulfanyl thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde,5-methylthio, 2-formyl-5-methylthiothiphene, 2-formyl-5-methylthio thiophene, 5-methylmercapto-2-thiophenaldehyde, 5-methylthio-2-thiophenecarbaldehyde, 5-methylthiothiophene-2-carbaldehyde, 5-methylmercaptothiophene-2-carbaldehyde
PubChem CID 2736997
IUPAC Name 5-methylsulfanylthiophene-2-carbaldehyde
SMILES CSC1=CC=C(S1)C=O

Specifications

Melting Point 26°C
Color White-Yellow
Boiling Point 95°C
Quantity 1 g
Formula Weight 158.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-(Methylthio)thiophene-2-carboxaldehyde
TSCA No
Recommended Storage Refrigerator