missing translation for 'onlineSavingsMsg'
Learn More

5-Methylsalicylic Acid 98.0+%, TCI America™

Catalog No. C04105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C04105G 5 g
C041025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C04105G Supplier TCI America Supplier No. C04105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 89-56-5
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002461
InChI Key DLGBEGBHXSAQOC-UHFFFAOYSA-N
Synonym 5-methylsalicylic acid, p-cresotic acid, p-cresotinic acid, 6-hydroxy-m-toluic acid, p-homosalicylic acid, 2,5-cresotic acid, benzoic acid, 2-hydroxy-5-methyl, 5-methyl-2-hydroxybenzoic acid, 6-hydroxy-3-methylbenzoic acid, alpha-cresotinic acid
PubChem CID 6973
IUPAC Name 2-hydroxy-5-methylbenzoic acid
SMILES CC1=CC(=C(C=C1)O)C(=O)O

Specifications

Melting Point 151°C
Color White-Yellow
Quantity 5 g
Formula Weight 152.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylsalicylic Acid
RTECSNumber GP3920200
TSCA No