missing translation for 'onlineSavingsMsg'
Learn More

5-Methylpyrazine-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. M10035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10035G 5 g
M100325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10035G Supplier TCI America Supplier No. M10035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5521-55-1
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.126
MDL Number MFCD00068241
InChI Key RBYJWCRKFLGNDB-UHFFFAOYSA-N
Synonym 5-methyl-2-pyrazinecarboxylic acid, 2-methylpyrazine-5-carboxylic acid, pyrazinecarboxylic acid, 5-methyl, unii-b861rs5nhi, 5-methylpyrazinecarboxylic acid, 5-methyl pyrazine-2-carboxylic acid, 5-methyl-pyrazine-2-carboxylic acid, 2-carboxy-5-methylpyrazine, 2-methyl-5-pyrazinoic acid, b861rs5nhi
PubChem CID 122831
IUPAC Name 5-methylpyrazine-2-carboxylic acid
SMILES CC1=NC=C(N=C1)C(=O)O

Specifications

Melting Point 166°C
Quantity 5 g
Formula Weight 138.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylpyrazine-2-carboxylic Acid