missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole 95.0+%, TCI America™

Catalog No. M08135ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08135ML 5 mL
M081325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08135ML Supplier TCI America Supplier No. M08135ML
Only null left

Chemical Identifiers

CAS 5765-44-6
Molecular Formula C4H5NO
Molecular Weight (g/mol) 83.09
MDL Number MFCD00003152
InChI Key AGQOIYCTCOEHGR-UHFFFAOYSA-N
Synonym 5-methylisoxazole, isoxazole, 5-methyl, unii-yc2p4ibf1e, yc2p4ibf1e, 5-methyl isoxazole, 5-methylisoxazol, 5-methyl-isoxazole, pubchem8634, acmc-1arbg
PubChem CID 79833
IUPAC Name 5-methyl-1,2-oxazole
SMILES CC1=CC=NO1

Specifications

Color Yellow
Boiling Point 122°C
Quantity 5 mL
UN Number 1993
Formula Weight 83.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Methylisoxazole