missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisoxazole-4-carboxylic Acid 98.0+%, TCI America™

Catalog No. M17405G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M17405G 5 g
M174025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M17405G Supplier TCI America Supplier No. M17405G

Chemical Identifiers

CAS 42831-50-5
Molecular Formula C5H4NO3
Molecular Weight (g/mol) 126.09
MDL Number MFCD00955655
InChI Key VQBXUKGMJCPBMF-UHFFFAOYSA-M
Synonym 5-methylisoxazole-4-carboxylic acid, 5-methyl-4-isoxazolecarboxylic acid, unii-bl2q213q2m, 4-isoxazolecarboxylic acid, 5-methyl, 5-methylisoxazole-4-carboxylicacid, leflunomide impurity d ep, pubchem8652, leflunomide impurity d, acmc-209jr8, ksc235o3b
PubChem CID 1425240
IUPAC Name 5-methyl-1,2-oxazole-4-carboxylate
SMILES CC1=C(C=NO1)C([O-])=O

Specifications

Melting Point 147°C
Color White-Yellow
Quantity 5 g
Formula Weight 127.10
Percent Purity ≥98.0% (T,HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylisoxazole-4-carboxylic Acid