missing translation for 'onlineSavingsMsg'
Learn More

5-Methylisophthalic Acid 98.0+%, TCI America™

Catalog No. M19035G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19035G 5 g
M190325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M19035G Supplier TCI America Supplier No. M19035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 499-49-0
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00013986
InChI Key PMZBHPUNQNKBOA-UHFFFAOYSA-N
Synonym 5-methylisophthalic acid, uvitic acid, 5-methyl-1,3-benzenedicarboxylic acid, isophthalic acid, 5-methyl, 1,3-benzenedicarboxylic acid, 5-methyl, uvitinsaure, 3,5-dicarboxytoluene, 5-methylisophthalicacid, acmc-209kj2, ksc237e4t
PubChem CID 68137
IUPAC Name 5-methylbenzene-1,3-dicarboxylic acid
SMILES CC1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 298°C
Quantity 5 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methylisophthalic Acid
EINECSNumber (3)-1373
TSCA No